Gene Transcripts

Gene Symbol
COA3
Entrez Gene ID
28958
Gene Name
cytochrome c oxidase assembly factor 3
Gene Aliases
CCDC56, HSPC009, MITRAC12
Chromosome Location
Chr. 17: 42797618 - 42798725 on Build GRCh38
UniGene ID
Hs.16059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040431.2 NP_001035521.1 2 762
GenBank mRNA AF070665.1 - 2 737
AK225611.1 - 2 741
BC002698.2 - 2 723 3609536
L38939.1 - 1 473

View Full Product Details