Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014049.4 | NP_054768.2 | 7 | 941 | |
NR_033426.1 | 7 | 1117 | |||
XM_017006264.1 | XP_016861753.1 | 7 | 1105 | ||
XM_017006265.1 | XP_016861754.1 | 1 | 87 | ||
GenBank mRNA | AB209427.1 | - | 7 | 2634 | |
AF327351.1 | - | 7 | 771 | ||
AK022568.1 | - | 6 | 867 | ||
AK024012.1 | - | 7 | 785 | ||
AK130311.1 | - | 7 | 812 | ||
AK294597.1 | - | 7 | 609 | ||
AK297952.1 | - | 7 | 985 | ||
AK310644.1 | - | 6 | 714 | ||
BC007970.2 | - | 7 | 818 | 4304209 | |
BC013354.2 | - | 7 | 795 | 3935925 |