Gene Transcripts
- Gene Symbol
- FLVCR1
- Entrez Gene ID
- 28982
- Gene Name
- feline leukemia virus subgroup C cellular receptor 1
- Gene Aliases
- AXPC1, FLVCR, MFSD7B, PCA, PCARP
- Chromosome Location
- Chr. 1: 212858255 - 212899363 on Build GRCh38
- UniGene ID
- Hs.7055
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF118637.1 |
- |
5, 6 |
1404 |
|
View Full Product Details