Gene Transcripts
- Gene Symbol
- MRPL22
- Entrez Gene ID
- 29093
- Gene Name
- mitochondrial ribosomal protein L22
- Gene Aliases
- HSPC158, L22mt, MRP-L22, MRP-L25, RPML25
- Chromosome Location
- Chr. 5: 154941073 - 154969411 on Build GRCh38
- UniGene ID
- Hs.483924
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001014990.2 |
NP_001014990.1
|
5, 6 |
323 |
|
NM_014180.3 |
NP_054899.2
|
6, 7 |
441 |
|
GenBank mRNA |
AF161507.1 |
- |
6, 7 |
409 |
|
AK000601.1 |
- |
6, 7 |
428 |
|
AK097880.1 |
- |
2, 3 |
2078 |
|
AK302784.1 |
- |
5, 6 |
303 |
|
BC012565.1 |
- |
6, 7 |
411 |
4333117 |
BP372420.1 |
- |
6, 7 |
441 |
|
BU943770.1 |
- |
6, 7 |
434 |
|
CR933679.1 |
- |
5, 6 |
302 |
|
View Full Product Details