Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013353.2 | NP_037485.2 | 8 | 965 | |
XM_011509449.1 | XP_011507751.1 | 8 | 847 | ||
XM_017001089.1 | XP_016856578.1 | 7 | 1582 | ||
GenBank mRNA | AF165217.1 | - | 8 | 842 | |
AF177173.2 | - | 8 | 910 | ||
AK092700.1 | - | 8 | 827 | ||
AK300675.1 | - | 6 | 668 | ||
AK313280.1 | - | 8 | 874 | ||
BC017810.1 | - | 8 | 875 | 4288769 | |
BX647346.1 | - | 6 | 589 |