Gene Transcripts

Gene Symbol
ZDHHC1
Entrez Gene ID
29800
Gene Name
zinc finger DHHC-type containing 1
Gene Aliases
C16orf1, HSU90653, ZNF377
Chromosome Location
Chr. 16: 67394419 - 67416494 on Build GRCh38
UniGene ID
Hs.624541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013304.2 NP_037436.1 10 1398
XM_011523060.1 XP_011521362.1 8 931
GenBank mRNA BC021908.1 - 10 1424

View Full Product Details