Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015971.3 | NP_057055.2 | 4 | 642 | |
GenBank mRNA | AF077042.1 | - | 4 | 591 | |
AK222995.1 | - | 4 | 606 | ||
AK313854.1 | - | 4 | 416 | ||
BC000241.1 | - | 4 | 601 | 3350465 | |
BC050641.2 | - | 4 | 398 | ||
BU599633.1 | - | 4 | 620 |