Gene Transcripts

Gene Symbol
SAR1B
Entrez Gene ID
51128
Gene Name
secretion associated Ras related GTPase 1B
Gene Aliases
ANDD, CMRD, GTBPB, SARA2
Chromosome Location
Chr. 5: 134601149 - 134632843 on Build GRCh38
UniGene ID
Hs.432984
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033503.2 NP_001028675.1 4 335
NM_016103.3 NP_057187.1 3 216
XM_017009521.1 XP_016865010.1 3 223
GenBank mRNA AF087850.1 - 3 253
AF092130.1 - 3 232
AK223452.1 - 3 201
AL710660.1 - 4 317
BC002847.2 - 4 266 3636540
BC093034.1 - 3 216 30528518
CD048852.1 - 4 343

View Full Product Details