Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320327.1 | NP_001307256.1 | 3 | 562 | |
NM_016139.3 | NP_057223.1 | 3 | 562 | ||
GenBank mRNA | AF078845.1 | - | 3 | 426 | |
AY605046.1 | - | 3 | 345 | ||
AY633613.1 | - | 3 | 366 | ||
BC003079.2 | - | 3 | 382 | 2988156 | |
BC015639.2 | - | 3 | 366 | 4637353 | |
BC066331.1 | - | 3 | 385 | 4792941 | |
BC071985.1 | - | 3 | 382 | 6426679 | |
BC100275.1 | - | 3 | 378 | 6192342 | |
BF347122.1 | - | 437 | |||
CN363208.1 | - | 3 | 402 |