Gene Transcripts

Gene Symbol
CDC25C
Entrez Gene ID
995
Gene Name
cell division cycle 25C
Gene Aliases
CDC25, PPP1R60
Chromosome Location
Chr. 5: 138285265 - 138338355 on Build GRCh38
UniGene ID
Hs.656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287582.1 NP_001274511.1 2 438
NM_001287583.1 NP_001274512.1 2 555
NM_001318098.1 NP_001305027.1 2 559
NM_001790.4 NP_001781.2 2 442
NM_022809.3 NP_073720.1 2 442
XM_005272145.3 XP_005272202.1 2 555
XM_011543759.2 XP_011542061.1 3 226
XM_011543760.2 XP_011542062.1 2 555
XM_011543761.2 XP_011542063.1 2 555
XM_011543762.1 XP_011542064.1 2 355
XM_011543764.1 XP_011542066.1 1 28
XM_017010101.1 XP_016865590.1 2 433
GenBank mRNA AF277723.1 - 1 28
AF277724.1 - 1 28
AF277725.1 - 1 28
AF277726.1 - 1 28
AJ304504.1 - 1 121
AK097710.1 - 2 555
AK301828.1 - 3 505
AU125025.1 - 2 363
BC019089.2 - 2 332 4858949
DB040755.1 - 2 363
DB449713.1 - 2 436
M34065.1 - 2 331

View Full Product Details