Gene Transcripts

Gene Symbol
WDR83OS
Entrez Gene ID
51398
Gene Name
WD repeat domain 83 opposite strand
Gene Aliases
C19orf56, PTD008
Chromosome Location
Chr. 19: 12668067 - 12671023 on Build GRCh38
UniGene ID
Hs.108969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016145.3 NP_057229.1 3 434
XM_017026864.1 XP_016882353.1 4 1428
GenBank mRNA AF059620.1 - 3 202
AF078861.1 - 3 419
AF151898.1 - 3 418
AK091030.1 - 3 2139
AK311944.1 - 3 198
BC001192.1 - 3 182
BC042919.1 - 3 203 5932109
BC061912.1 - 3 203 4444565

View Full Product Details