Gene Transcripts
- Gene Symbol
- UFM1
- Entrez Gene ID
- 51569
- Gene Name
- ubiquitin fold modifier 1
- Gene Aliases
- BM-002, C13orf20
- Chromosome Location
- Chr. 13: 38349771 - 38363007 on Build GRCh38
- UniGene ID
- Hs.643655
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286703.1 |
NP_001273632.1
|
5, 6 |
332 |
|
NM_001286704.1 |
NP_001273633.1
|
5, 6 |
510 |
|
NM_016617.3 |
NP_057701.1
|
5, 6 |
338 |
|
NR_104584.1 |
|
6, 7 |
399 |
|
NR_104585.1 |
|
4, 5 |
298 |
|
GenBank mRNA |
AB154404.1 |
- |
4, 5 |
189 |
|
AF208844.1 |
- |
5, 6 |
228 |
|
AK095490.1 |
- |
5, 6 |
258 |
|
AL515797.3 |
- |
4, 5 |
190 |
|
BC005193.1 |
- |
5, 6 |
255 |
3829206 |
BI458294.1 |
- |
5, 6 |
309 |
|
BI761948.1 |
- |
5, 6 |
215 |
|
BP277186.1 |
- |
5, 6 |
338 |
|
BU589105.1 |
- |
6, 7 |
315 |
|
CR457189.1 |
- |
4, 5 |
189 |
|
DA448931.1 |
- |
4, 5 |
211 |
|
DA483023.1 |
- |
5, 6 |
258 |
|
Z70222.1 |
- |
5, 6 |
229 |
|
View Full Product Details