Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016930.2 | NP_058626.1 | 4 | 484 | |
XM_011513480.1 | XP_011511782.1 | 4 | 463 | ||
XM_011513483.1 | XP_011511785.1 | 4 | 463 | ||
XM_011513485.2 | XP_011511787.1 | 4 | 411 | ||
XM_017008296.1 | XP_016863785.1 | 4 | 463 | ||
XM_017008297.1 | XP_016863786.1 | 4 | 1044 | ||
XM_017008298.1 | XP_016863787.1 | 4 | 463 | ||
GenBank mRNA | AB028741.1 | - | 4 | 403 | |
AK310026.1 | - | 4 | 446 | ||
AY453396.1 | - | 4 | 400 | ||
BC014613.2 | - | 4 | 406 | 2960768 | |
BG771856.1 | - | 4 | 489 | ||
BT007150.1 | - | 4 | 400 | ||
BT020135.1 | - | 4 | 400 |