Gene Transcripts

Gene Symbol
CSNK1G1
Entrez Gene ID
53944
Gene Name
casein kinase 1 gamma 1
Gene Aliases
CK1gamma1
Chromosome Location
Chr. 15: 64165517 - 64356243 on Build GRCh38
UniGene ID
Hs.646508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022048.3 NP_071331.2 4 735
GenBank mRNA AB042562.1 - 3 383
AB042563.1 - 3 367
AF223357.2 - 3 453
AK225960.1 - 4 665
AL832551.1 - 4 513
BC017236.2 - 3 184
BC130613.1 - 4 508 40146644
BC136504.1 - 4 508 9020493
CF593592.1 - 4 693

View Full Product Details