Gene Transcripts

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
Chromosome Location
Chr. 9: 136940433 - 136944774 on Build GRCh38
UniGene ID
Hs.522507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018998.3 NP_061871.1 7 1377
XM_005266089.2 XP_005266146.2 7 1427
XM_011518789.1 XP_011517091.1 7 1373
XM_017014812.1 XP_016870301.1 7 1343
GenBank mRNA AB209726.1 - 4 789
AF217998.1 - 3 537
AF258569.1 - 3 535
AK026081.1 - 7 1241
AK315738.1 - 7 1252
AL137631.1 - 3 931
BC004541.2 - 4 786
BC009429.1 - 2 530
BC014130.1 - 7 1273 4620371
BC014297.2 - 7 1244 3958395
BC031100.1 - 6 1338
BC035355.1 - 6 1391

View Full Product Details