Gene Transcripts
- Gene Symbol
- MXRA8
- Entrez Gene ID
- 54587
- Gene Name
- matrix remodeling associated 8
- Gene Aliases
- Chromosome Location
- Chr. 1: 1352689 - 1363541 on Build GRCh38
- UniGene ID
- Hs.515687
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282582.1 |
NP_001269511.1
|
9, 10 |
1288 |
|
NM_001282583.1 |
NP_001269512.1
|
8, 9 |
1707 |
|
NM_001282584.1 |
NP_001269513.1
|
7, 8 |
1207 |
|
NM_032348.3 |
NP_115724.1
|
8, 9 |
1510 |
|
XM_017001516.1 |
XP_016857005.1
|
8, 9 |
1287 |
|
XM_017001517.1 |
XP_016857006.1
|
7, 8 |
2914 |
|
GenBank mRNA |
AB052096.1 |
- |
9, 10 |
1288 |
|
AK095966.1 |
- |
8, 9 |
1707 |
|
AK225028.1 |
- |
8, 9 |
1249 |
|
AK293442.1 |
- |
7, 8 |
976 |
|
AK304615.1 |
- |
7, 8 |
1302 |
|
AK315510.1 |
- |
8, 9 |
1277 |
|
AL050202.1 |
- |
2, 3 |
146 |
|
AY358391.1 |
- |
7, 8 |
953 |
|
BC006213.2 |
- |
8, 9 |
1251 |
3343261 |
BC017312.2 |
- |
8, 9 |
1251 |
5090754 |
View Full Product Details