Gene Transcripts

Gene Symbol
ZNHIT6
Entrez Gene ID
54680
Gene Name
zinc finger HIT-type containing 6
Gene Aliases
BCD1, C1orf181, NY-BR-75
Chromosome Location
Chr. 1: 85649423 - 85708447 on Build GRCh38
UniGene ID
Hs.5111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170670.1 NP_001164141.1 4, 5 862
NM_017953.3 NP_060423.3 3, 4 979
XM_011541614.2 XP_011539916.1 3, 4 993
GenBank mRNA AF308296.1 - 3, 4 626
AK000736.1 - 3, 4 351
AK000767.1 - 3, 4 888
AK298149.1 - 4, 5 862
AK314566.1 - 3, 4 907
AL442074.1 - 3, 4 964
BC026236.1 - 3, 4 966 4664103
BC110898.1 - 3, 4 897 6012059

View Full Product Details