Gene Transcripts

Gene Symbol
FAM46C
Entrez Gene ID
54855
Gene Name
family with sequence similarity 46 member C
Gene Aliases
Chromosome Location
Chr. 1: 117605982 - 117628389 on Build GRCh38
UniGene ID
Hs.356216
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017709.3 NP_060179.2 2 800
GenBank mRNA AK000209.1 - 2 735
BC036516.2 - 2 729 5267660
BC131726.1 - 2 713 40123993

View Full Product Details