Gene Transcripts
- Gene Symbol
- ST7L
- Entrez Gene ID
- 54879
- Gene Name
- suppression of tumorigenicity 7 like
- Gene Aliases
- FAM4B, ST7R, STLR
- Chromosome Location
- Chr. 1: 112517444 - 112620838 on Build GRCh38
- UniGene ID
- Hs.201921
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017744.4 |
NP_060214.2
|
4, 5 |
809 |
|
NM_138727.3 |
NP_620055.1
|
4, 5 |
758 |
|
NM_138728.2 |
NP_620056.1
|
4, 5 |
809 |
|
NM_138729.3 |
NP_620057.1
|
4, 5 |
809 |
|
XM_005270963.4 |
XP_005271020.1
|
4, 5 |
332 |
|
XM_005270964.3 |
XP_005271021.1
|
3, 4 |
424 |
|
XM_011541627.2 |
XP_011539929.1
|
4, 5 |
518 |
|
XM_011541628.2 |
XP_011539930.1
|
4, 5 |
518 |
|
XM_011541629.2 |
XP_011539931.1
|
4, 5 |
532 |
|
XM_011541630.2 |
XP_011539932.1
|
4, 5 |
1228 |
|
XM_011541631.2 |
XP_011539933.1
|
4, 5 |
356 |
|
XM_017001534.1 |
XP_016857023.1
|
4, 5 |
518 |
|
XM_017001535.1 |
XP_016857024.1
|
4, 5 |
518 |
|
XM_017001536.1 |
XP_016857025.1
|
3, 4 |
424 |
|
XM_017001538.1 |
XP_016857027.1
|
3, 4 |
299 |
|
GenBank mRNA |
AB081317.1 |
- |
4, 5 |
580 |
|
AB081318.1 |
- |
4, 5 |
529 |
|
AB081319.1 |
- |
4, 5 |
580 |
|
AB081320.1 |
- |
4, 5 |
580 |
|
AK021719.1 |
- |
3, 4 |
419 |
|
AK095837.1 |
- |
2, 3 |
117 |
|
AK291042.1 |
- |
4, 5 |
510 |
|
AK301869.1 |
- |
4, 5 |
510 |
|
AK304004.1 |
- |
4, 5 |
467 |
|
AK308727.1 |
- |
3, 4 |
433 |
|
BC027898.1 |
- |
3, 4 |
411 |
|
BC037830.1 |
- |
4, 5 |
1222 |
4812133 |
BC047392.1 |
- |
4, 5 |
535 |
5273549 |
BC065501.1 |
- |
4, 5 |
716 |
4417786 |
BX537988.1 |
- |
5, 6 |
671 |
|
BX640667.1 |
- |
4, 5 |
485 |
|
View Full Product Details