Gene Transcripts

Gene Symbol
UHRF1BP1
Entrez Gene ID
54887
Gene Name
UHRF1 binding protein 1
Gene Aliases
C6orf107, ICBP90, dJ349A12.1
Chromosome Location
Chr. 6: 34792017 - 34877514 on Build GRCh38
UniGene ID
Hs.700656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017754.3 NP_060224.3 13 1861
GenBank mRNA AB126777.1 - 13 1696
BC137120.1 - 13 1691 9021117
BC137127.1 - 13 1691 9021124
BX640641.1 - 13 1730

View Full Product Details