Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001305544.1 | NP_001292473.1 | 2 | 861 | |
NM_017763.5 | NP_060233.3 | 2 | 1064 | ||
XM_011524955.2 | XP_011523257.1 | 2 | 963 | ||
XM_017024800.1 | XP_016880289.1 | 3 | 2290 | ||
GenBank mRNA | AB081837.1 | - | 2 | 601 | |
AK000322.1 | - | 1 | 281 | ||
AK291027.1 | - | 2 | 834 | ||
AK296769.1 | - | 1 | 177 | ||
BC109028.2 | - | 1 | 310 | 40027722 | |
CR627423.1 | - | 2 | 861 | ||
CR749257.1 | - | 1 | 2069 |