Gene Transcripts
- Gene Symbol
- RHBDL2
- Entrez Gene ID
- 54933
- Gene Name
- rhomboid like 2
- Gene Aliases
- RRP2
- Chromosome Location
- Chr. 1: 38885806 - 38941830 on Build GRCh38
- UniGene ID
- Hs.524626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304746.1 |
NP_001291675.1
|
2, 3 |
501 |
|
NM_017821.4 |
NP_060291.2
|
2, 3 |
531 |
|
GenBank mRNA |
AK097621.1 |
- |
2, 3 |
1266 |
|
AK294115.1 |
- |
2, 3 |
501 |
|
AY126343.1 |
- |
1, 2 |
257 |
|
BC013103.1 |
- |
2, 3 |
502 |
|
BC137108.1 |
- |
1, 2 |
380 |
9021105 |
BC137110.1 |
- |
1, 2 |
380 |
9021107 |
BC144476.1 |
- |
1, 2 |
380 |
|
DB036310.1 |
- |
2, 3 |
531 |
|
View Full Product Details