Gene Transcripts

Gene Symbol
OXR1
Entrez Gene ID
55074
Gene Name
oxidation resistance 1
Gene Aliases
Nbla00307, TLDC3
Chromosome Location
Chr. 8: 106270178 - 106752694 on Build GRCh38
UniGene ID
Hs.127286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198532.1 NP_001185461.1 4 425
NM_001198533.1 NP_001185462.1 5 651
NM_018002.3 NP_060472.2 5 651
NM_181354.4 NP_851999.2 3 481
XM_006716595.2 XP_006716658.1 4 420
XM_017013589.1 XP_016869078.1 3 450
XM_017013590.1 XP_016869079.1 3 291
XM_017013591.1 XP_016869080.1 3 381
XM_017013592.1 XP_016869081.1 3 128
XM_017013593.1 XP_016869082.1 3 355
XM_017013594.1 XP_016869083.1 3 355
XM_017013595.1 XP_016869084.1 3 450
XM_017013596.1 XP_016869085.1 5 623
XM_017013597.1 XP_016869086.1 3 284
XM_017013598.1 XP_016869087.1 3 355
XM_017013599.1 XP_016869088.1 3 450
XM_017013600.1 XP_016869089.1 3 307
GenBank mRNA AK096148.1 - 3 161
AK124441.1 - 5 584
AK292899.1 - 3 433
AK296561.1 - 4 425
AL833193.1 - 2 62
BC019691.1 - 3 298
BC032710.2 - 3 423 5496492
BC035484.1 - 3 435
FN650108.1 - 4 461

View Full Product Details