Gene Transcripts

Gene Symbol
SLC47A1
Entrez Gene ID
55244
Gene Name
solute carrier family 47 member 1
Gene Aliases
MATE1
Chromosome Location
Chr. 17: 19533854 - 19579033 on Build GRCh38
UniGene ID
Hs.232054
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018242.2 NP_060712.2 7 655
GenBank mRNA AK001709.1 - 7 655
AK222625.1 - 7 659
AK301854.1 - 6 612
BC010661.2 - 7 644 3874616
BC050592.1 - 7 629
BC058882.1 - 7 631 5188651

View Full Product Details