Gene Transcripts

Gene Symbol
LRRC36
Entrez Gene ID
55282
Gene Name
leucine rich repeat containing 36
Gene Aliases
RORBP70, XLHSRF2
Chromosome Location
Chr. 16: 67326844 - 67385206 on Build GRCh38
UniGene ID
Hs.125139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161575.1 NP_001155047.1 11 1809
NM_018296.5 NP_060766.5 14 2079
XM_005256025.2 XP_005256082.1 13 1964
XM_005256026.2 XP_005256083.1 13 1638
XM_005256027.2 XP_005256084.1 12 1818
XM_005256028.1 XP_005256085.1 13 1951
XM_011523201.1 XP_011521503.1 11 1699
XM_011523202.2 XP_011521504.1 10 1554
XM_011523203.1 XP_011521505.1 11 1461
XM_011523204.1 XP_011521506.1 12 1878
XM_011523205.1 XP_011521507.1 13 1969
XM_017023401.1 XP_016878890.1 11 1182
XM_017023402.1 XP_016878891.1 9 1005
GenBank mRNA AK001866.1 - 10 1712
AK292311.1 - 11 1788
AK297329.1 - 8 1146
AK301766.1 - 9 1451
AY313780.1 - 11 1772
AY963267.1 - 14 2079
BC018031.1 - 13 1573
BC026156.2 - 14 2079 4828487

View Full Product Details