Gene Transcripts
- Gene Symbol
- ABCF3
- Entrez Gene ID
- 55324
- Gene Name
- ATP binding cassette subfamily F member 3
- Gene Aliases
- EST201864
- Chromosome Location
- Chr. 3: 184186075 - 184194012 on Build GRCh38
- UniGene ID
- Hs.361323
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018358.2 |
NP_060828.2
|
4, 5 |
485 |
|
XM_011512959.2 |
XP_011511261.1
|
4, 5 |
489 |
|
GenBank mRNA |
AK002060.1 |
- |
4, 5 |
462 |
|
AK027288.1 |
- |
4, 5 |
360 |
|
AK092415.1 |
- |
4, 5 |
368 |
|
AK290106.1 |
- |
4, 5 |
357 |
|
AK299256.1 |
- |
4, 5 |
364 |
|
AK299433.1 |
- |
4, 5 |
249 |
|
BC009253.2 |
- |
4, 5 |
364 |
3028775 |
BC035652.1 |
- |
4, 5 |
359 |
|
BC051754.2 |
- |
4, 5 |
359 |
6064628 |
BC051884.1 |
- |
4, 5 |
359 |
5105761 |
DB079392.1 |
- |
4, 5 |
417 |
|
DC318082.1 |
- |
4, 5 |
537 |
|
KU178694.1 |
- |
4, 5 |
352 |
|
KU178695.1 |
- |
4, 5 |
352 |
|
View Full Product Details