Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252148.1 | NP_001239077.1 | 5 | 1242 | |
NM_001252150.1 | NP_001239079.1 | 6 | 1311 | ||
NM_001252151.1 | NP_001239080.1 | 6 | 738 | ||
NM_001252152.1 | NP_001239081.1 | 7 | 726 | ||
NM_018375.4 | NP_060845.2 | 6 | 1311 | ||
XM_005267834.1 | XP_005267891.1 | 5 | 1113 | ||
XM_017021426.1 | XP_016876915.1 | 7 | 1452 | ||
GenBank mRNA | AK002136.1 | - | 6 | 1287 | |
AK222547.1 | - | 6 | 1289 | ||
AK293222.1 | - | 6 | 731 | ||
AK316381.1 | - | 7 | 715 | ||
AY358687.1 | - | 6 | 757 | ||
AY780789.1 | - | 5 | 540 | ||
BC047682.1 | - | 6 | 1241 | 6066186 | |
BC064383.1 | - | 6 | 1026 | 5737339 | |
BP269907.1 | - | 5 | 513 | ||
FJ423549.1 | - | 6 | 609 |