Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
55353
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
LAPTM4beta, LC27
Chromosome Location
Chr. 8: 97775581 - 97852602 on Build GRCh38
UniGene ID
Hs.492314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018407.4 NP_060877.3 2 587
GenBank mRNA AF317417.1 - 2 158
AF527412.1 - 2 516
AK075326.1 - 2 349
AL136942.1 - 2 341
AY057051.2 - 2 587
AY261384.1 - 2 606
BC014129.2 - 2 343 4581284
BC031021.1 - 2 368 4690368
BC038117.1 - 2 1135 5264567

View Full Product Details