Gene Transcripts

Gene Symbol
G2E3
Entrez Gene ID
55632
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
Gene Aliases
KIAA1333, PHF7B
Chromosome Location
Chr. 14: 30559123 - 30620064 on Build GRCh38
UniGene ID
Hs.509008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308097.1 NP_001295026.1 13, 14 1985
NM_017769.4 NP_060239.2 14, 15 2026
XM_011536925.2 XP_011535227.1 14, 15 1992
XM_011536926.1 XP_011535228.1 13, 14 1909
GenBank mRNA AB037754.1 - 14, 15 1978
AK000340.1 - 14, 15 1991
AK225346.1 - 14, 15 1991
AK296872.1 - 13, 14 1950
BC000973.2 - 14, 15 1972 3450488
BC040179.1 - 2, 3 1282

View Full Product Details