Gene Transcripts

Gene Symbol
ZC4H2
Entrez Gene ID
55906
Gene Name
zinc finger C4H2-type containing
Gene Aliases
HCA127, KIAA1166, WRWF, WWS
Chromosome Location
Chr. X: 64915802 - 65034744 on Build GRCh38
UniGene ID
Hs.28249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178032.2 NP_001171503.1 3, 4 713
NM_001243804.1 NP_001230733.1 3, 4 458
NM_018684.3 NP_061154.1 3, 4 551
NR_045044.1 3, 4 806
GenBank mRNA AF270491.1 - 3, 4 483
AK022807.1 - 3, 4 682
AK022918.1 - 3, 4 445
AK123763.1 - 4, 5 944
AK222708.1 - 3, 4 468
AK315485.1 - 3, 4 466
BC004411.1 - 3, 4 502 3639978
BI770655.1 - 3, 4 636
BP376634.1 - 3, 4 458
CX782941.1 - 3, 4 423

View Full Product Details