Gene Transcripts
- Gene Symbol
- LANCL2
- Entrez Gene ID
- 55915
- Gene Name
- LanC like 2
- Gene Aliases
- GPR69B, TASP
- Chromosome Location
- Chr. 7: 55365448 - 55433742 on Build GRCh38
- UniGene ID
- Hs.595384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018697.3 |
NP_061167.1
|
7, 8 |
1756 |
|
GenBank mRNA |
AB035966.1 |
- |
7, 8 |
1233 |
|
AF353942.1 |
- |
7, 8 |
1233 |
|
AJ278245.1 |
- |
7, 8 |
1364 |
|
AK095857.1 |
- |
7, 8 |
1756 |
|
AK225958.1 |
- |
7, 8 |
1209 |
|
AK301747.1 |
- |
7, 8 |
1162 |
|
AK313326.1 |
- |
7, 8 |
1501 |
|
AL713665.2 |
- |
7, 8 |
1148 |
|
BC004887.2 |
- |
4, 5 |
584 |
|
BC070049.1 |
- |
7, 8 |
1452 |
30343188 |
View Full Product Details