Gene Transcripts
- Gene Symbol
- SLC50A1
- Entrez Gene ID
- 55974
- Gene Name
- solute carrier family 50 member 1
- Gene Aliases
- HsSWEET1, RAG1AP1, SCP, SWEET1, slv
- Chromosome Location
- Chr. 1: 155135253 - 155138858 on Build GRCh38
- UniGene ID
- Hs.292154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122837.1 |
NP_001116309.1
|
3, 4 |
430 |
|
NM_001122839.1 |
NP_001116311.1
|
3, 4 |
392 |
|
NM_001287586.1 |
NP_001274515.1
|
3, 4 |
457 |
|
NM_001287587.1 |
NP_001274516.1
|
4, 5 |
574 |
|
NM_001287588.1 |
NP_001274517.1
|
3, 4 |
430 |
|
NM_001287590.1 |
NP_001274519.1
|
3, 4 |
587 |
|
NM_001287591.1 |
NP_001274520.1
|
4, 5 |
611 |
|
NM_018845.3 |
NP_061333.2
|
4, 5 |
554 |
|
XM_005245339.4 |
XP_005245396.1
|
4, 5 |
740 |
|
XM_005245340.3 |
XP_005245397.1
|
4, 5 |
821 |
|
XM_005245341.4 |
XP_005245398.1
|
4, 5 |
772 |
|
XM_005245342.3 |
XP_005245399.1
|
3, 4 |
380 |
|
XM_006711453.2 |
XP_006711516.1
|
4, 5 |
554 |
|
GenBank mRNA |
AF126023.1 |
- |
4, 5 |
514 |
|
AF126024.1 |
- |
3, 4 |
379 |
|
AK289551.1 |
- |
4, 5 |
555 |
|
BC005943.1 |
- |
4, 5 |
498 |
4076256 |
BC009621.1 |
- |
3, 4 |
354 |
3896154 |
BC010869.1 |
- |
3, 4 |
345 |
|
BC012378.1 |
- |
3, 4 |
379 |
|
BE958344.1 |
- |
|
411 |
|
BI195885.1 |
- |
3, 4 |
589 |
|
BM806886.1 |
- |
4, 5 |
598 |
|
BU160680.1 |
- |
4, 5 |
496 |
|
BU564973.1 |
- |
3, 4 |
417 |
|
CR457148.1 |
- |
4, 5 |
454 |
|
View Full Product Details