Gene Transcripts

Gene Symbol
SUCNR1
Entrez Gene ID
56670
Gene Name
succinate receptor 1
Gene Aliases
GPR91
Chromosome Location
Chr. 3: 151873643 - 151884619 on Build GRCh38
UniGene ID
Hs.133181
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033050.5 NP_149039.2 3 553
GenBank mRNA AF247785.1 - 3 516
AF348078.1 - 2 490
AK290420.1 - 3 553
BC030948.1 - 3 547 4594810
EU432110.1 - 1 448

View Full Product Details