Gene Transcripts
- Gene Symbol
- SLC12A5
- Entrez Gene ID
- 57468
- Gene Name
- solute carrier family 12 member 5
- Gene Aliases
- EIEE34, EIG14, KCC2, hKCC2
- Chromosome Location
- Chr. 20: 46021690 - 46060150 on Build GRCh38
- UniGene ID
- Hs.21413
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134771.1 |
NP_001128243.1
|
20, 21 |
2832 |
|
NM_020708.4 |
NP_065759.1
|
20, 21 |
2850 |
|
XM_017027981.1 |
XP_016883470.1
|
20, 21 |
2832 |
|
GenBank mRNA |
AB033002.1 |
- |
20, 21 |
2665 |
|
AF208159.1 |
- |
20, 21 |
2687 |
|
AK289758.1 |
- |
20, 21 |
2809 |
|
AK294059.1 |
- |
16, 17 |
2027 |
|
AK295096.1 |
- |
4, 5 |
392 |
|
AK295900.1 |
- |
14, 15 |
2046 |
|
BC132668.1 |
- |
20, 21 |
2775 |
40146690 |
BC132670.1 |
- |
20, 21 |
2775 |
40146692 |
View Full Product Details