Gene Transcripts
- Gene Symbol
- COG6
- Entrez Gene ID
- 57511
- Gene Name
- component of oligomeric golgi complex 6
- Gene Aliases
- CDG2L, COD2, SHNS
- Chromosome Location
- Chr. 13: 39655627 - 39791665 on Build GRCh38
- UniGene ID
- Hs.507805
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145079.1 |
NP_001138551.1
|
17, 18 |
1854 |
|
NM_020751.2 |
NP_065802.1
|
17, 18 |
1854 |
|
NR_026745.1 |
|
18, 19 |
1919 |
|
XM_011535168.1 |
XP_011533470.1
|
17, 18 |
1804 |
|
GenBank mRNA |
AB032960.1 |
- |
17, 18 |
1744 |
|
AF116827.1 |
- |
18, 19 |
1802 |
|
AK026638.1 |
- |
16, 17 |
1521 |
|
AK294443.1 |
- |
17, 18 |
1854 |
|
AK310953.1 |
- |
3, 4 |
345 |
|
BC051723.1 |
- |
17, 18 |
1804 |
5263056 |
CR627406.1 |
- |
17, 18 |
1797 |
|
View Full Product Details