Gene Transcripts

Gene Symbol
ARRDC3
Entrez Gene ID
57561
Gene Name
arrestin domain containing 3
Gene Aliases
TLIMP
Chromosome Location
Chr. 5: 91368724 - 91383359 on Build GRCh38
UniGene ID
Hs.24684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020801.2 NP_065852.1 1 420
GenBank mRNA AB037797.1 - 1 392
AK291753.1 - 1 395
BC015928.2 - 1 387 3908916
BC053619.1 - 1 392 6155815
DA110572.1 - 1 420

View Full Product Details