Gene Transcripts

Gene Symbol
PHRF1
Entrez Gene ID
57661
Gene Name
PHD and ring finger domains 1
Gene Aliases
PPP1R125, RNF221
Chromosome Location
Chr. 11: 576446 - 612225 on Build GRCh38
UniGene ID
Hs.325838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286581.1 NP_001273510.1 4 500
NM_001286582.1 NP_001273511.1 4 497
NM_001286583.1 NP_001273512.1 4 488
NM_020901.3 NP_065952.2 4 500
XM_005253025.4 XP_005253082.1 4 458
XM_005253027.3 XP_005253084.1 4 446
XM_011520236.2 XP_011518538.1 4 500
XM_011520237.2 XP_011518539.1 3 341
GenBank mRNA AB040975.1 - 4 460
BC112931.1 - 3 771
BC136615.1 - 4 425 9020605
BC144293.1 - 4 425 9052813
BC144294.1 - 4 413 9052814
BC144295.1 - 4 422 9052815
BC146771.1 - 4 460 9007218
DA989169.1 - 4 463

View Full Product Details