Gene Transcripts

Gene Symbol
SIGIRR
Entrez Gene ID
59307
Gene Name
single Ig and TIR domain containing
Gene Aliases
IL-1R8, TIR8
Chromosome Location
Chr. 11: 405716 - 417454 on Build GRCh38
UniGene ID
Hs.501624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135053.1 NP_001128525.1 7 936
NM_001135054.1 NP_001128526.1 7 1009
NM_021805.2 NP_068577.2 7 905
XM_005253044.2 XP_005253101.1 7 1009
XM_005253045.1 XP_005253102.1 7 905
XM_005253046.1 XP_005253103.1 7 993
XM_005253048.4 XP_005253105.1 5 699
XM_017018099.1 XP_016873588.1 7 1009
XM_017018100.1 XP_016873589.1 6 838
GenBank mRNA AK025099.1 - 7 905
AK093427.1 - 6 761
AK172830.1 - 7 905
AY358342.1 - 6 782
BC003591.1 - 6 835 2821373
BC025953.1 - 7 864 3939162
BC106007.1 - 7 986 6092503
BX369506.2 - 2 104
CR457338.1 - 5 679

View Full Product Details