Gene Transcripts
- Gene Symbol
- CDH26
- Entrez Gene ID
- 60437
- Gene Name
- cadherin 26
- Gene Aliases
- VR20
- Chromosome Location
- Chr. 20: 59957764 - 60035629 on Build GRCh38
- UniGene ID
- Hs.729046
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021810.4 |
NP_068582.2
|
4, 5 |
421 |
|
NM_177980.2 |
NP_817089.1
|
16, 17 |
2524 |
|
XM_011528969.2 |
XP_011527271.1
|
16, 17 |
2530 |
|
XM_011528970.2 |
XP_011527272.1
|
10, 11 |
1546 |
|
XM_017027994.1 |
XP_016883483.1
|
16, 17 |
3193 |
|
XM_017027995.1 |
XP_016883484.1
|
15, 16 |
3125 |
|
XM_017027996.1 |
XP_016883485.1
|
16, 17 |
3095 |
|
XM_017027998.1 |
XP_016883487.1
|
16, 17 |
3191 |
|
XM_017027999.1 |
XP_016883488.1
|
15, 16 |
3122 |
|
XM_017028000.1 |
XP_016883489.1
|
15, 16 |
2363 |
|
XM_017028002.1 |
XP_016883491.1
|
16, 17 |
3192 |
|
GenBank mRNA |
AK055202.1 |
- |
16, 17 |
2513 |
|
AK074477.1 |
- |
4, 5 |
409 |
|
BC136306.1 |
- |
16, 17 |
2507 |
9020293 |
BC143931.1 |
- |
16, 17 |
2507 |
|
View Full Product Details