Gene Transcripts

Gene Symbol
RTP4
Entrez Gene ID
64108
Gene Name
receptor transporter protein 4
Gene Aliases
IFRG28, Z3CXXC4
Chromosome Location
Chr. 3: 187368380 - 187371584 on Build GRCh38
UniGene ID
Hs.43388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022147.2 NP_071430.2 2 557
GenBank mRNA AJ251832.1 - 2 549
AY562238.1 - 2 495
BC013161.1 - 2 557 3919961

View Full Product Details