Gene Transcripts

Gene Symbol
CDH24
Entrez Gene ID
64403
Gene Name
cadherin 24
Gene Aliases
CDH11L
Chromosome Location
Chr. 14: 23047061 - 23057619 on Build GRCh38
UniGene ID
Hs.155912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022478.3 NP_071923.2 3 564
NM_144985.3 NP_659422.2 3 564
XM_011537089.1 XP_011535391.1 3 645
GenBank mRNA AK057922.1 - 3 500
AK289576.1 - 3 564
AK307746.1 - 3 546
AY260900.1 - 2 397
AY260901.1 - 2 397
AY358199.1 - 3 520
BC110867.1 - 3 535
BX248750.1 - 3 546

View Full Product Details