Gene Transcripts

Gene Symbol
GORASP1
Entrez Gene ID
64689
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
GOLPH5, GRASP65, P65
Chromosome Location
Chr. 3: 39096599 - 39108363 on Build GRCh38
UniGene ID
Hs.721946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278789.1 NP_001265718.1 6 1017
NM_001278790.1 NP_001265719.1 5 837
NM_031899.3 NP_114105.1 8 1765
NR_103866.1 8 1261
NR_103867.1 8 1253
XM_006713301.3 XP_006713364.1 8 1865
XM_011534020.2 XP_011532322.1 8 1775
XM_011534021.2 XP_011532323.1 7 1685
XM_017007050.1 XP_016862539.1 8 951
XM_017007051.1 XP_016862540.1 7 1300
XM_017007052.1 XP_016862541.1 7 896
GenBank mRNA AJ409349.1 - 8 1041
AK001574.1 - 7 1283
AK057043.1 - 5 575
AK124755.1 - 8 1765
AK303847.1 - 6 757
BC008928.2 - 8 982
BC075854.1 - 8 1051 5503263
BC103826.1 - 8 983 40001584
BC103827.1 - 8 983 40001588
BM470731.1 - 3 278

View Full Product Details