Gene Transcripts

Gene Symbol
TMEM237
Entrez Gene ID
65062
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4, JBTS14
Chromosome Location
Chr. 2: 201620184 - 201643529 on Build GRCh38
UniGene ID
Hs.12319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044385.2 NP_001037850.1 8 708
NM_152388.3 NP_689601.2 8 873
GenBank mRNA AB053301.1 - 8 875
AF212043.1 - 3 277
AK090601.1 - 8 566
AK303954.1 - 8 695
AK304395.1 - 6 397
BC029611.1 - 8 621 5304046
BQ219059.1 - 8 680

View Full Product Details