Gene Transcripts

Gene Symbol
YIPF2
Entrez Gene ID
78992
Gene Name
Yip1 domain family member 2
Gene Aliases
FinGER2
Chromosome Location
Chr. 19: 10922185 - 10929012 on Build GRCh38
UniGene ID
Hs.164026
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321439.1 NP_001308368.1 5 543
NM_001321440.1 NP_001308369.1 5 543
NM_024029.4 NP_076934.1 5 814
XM_011528270.2 XP_011526572.1 5 543
XM_011528271.2 XP_011526573.1 4 404
GenBank mRNA BC000056.2 - 5 462
BC013014.2 - 5 483 2960233
BM554546.1 - 5 461

View Full Product Details