Gene Transcripts

Gene Symbol
CCDC86
Entrez Gene ID
79080
Gene Name
coiled-coil domain containing 86
Gene Aliases
Chromosome Location
Chr. 11: 60841956 - 60851088 on Build GRCh38
UniGene ID
Hs.4253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024098.3 NP_077003.1 4 1157
GenBank mRNA AK025974.1 - 4 1043
AK123775.1 - 6 830
AK302329.1 - 5 947
BC001378.1 - 4 1008 3051044
BE795888.1 - 3 128
DQ501251.1 - 4 988

View Full Product Details