Gene Transcripts
- Gene Symbol
- GNPTAB
- Entrez Gene ID
- 79158
- Gene Name
- N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
- Gene Aliases
- GNPTA, ICD
- Chromosome Location
- Chr. 12: 101745497 - 101830867 on Build GRCh38
- UniGene ID
- Hs.46850
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024312.4 |
NP_077288.2
|
9, 10 |
1310 |
|
XM_006719593.2 |
XP_006719656.1
|
9, 10 |
1305 |
|
XM_011538731.2 |
XP_011537033.1
|
9, 10 |
1306 |
|
XM_017019961.1 |
XP_016875450.1
|
9, 10 |
1207 |
|
XM_017019962.1 |
XP_016875451.1
|
9, 10 |
1412 |
|
GenBank mRNA |
AB033034.2 |
- |
2, 3 |
201 |
|
AK056137.1 |
- |
9, 10 |
1181 |
|
AM085438.1 |
- |
9, 10 |
1297 |
|
AY687932.1 |
- |
9, 10 |
1282 |
|
BC071687.1 |
- |
9, 10 |
1258 |
|
BC131787.1 |
- |
9, 10 |
1238 |
40126580 |
View Full Product Details