Gene Transcripts

Gene Symbol
GNPTAB
Entrez Gene ID
79158
Gene Name
N-acetylglucosamine-1-phosphate transferase alpha and beta subunits
Gene Aliases
GNPTA, ICD
Chromosome Location
Chr. 12: 101745497 - 101830867 on Build GRCh38
UniGene ID
Hs.46850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024312.4 NP_077288.2 19 3757
XM_011538731.2 XP_011537033.1 19 3753
XM_017019961.1 XP_016875450.1 19 3654
XM_017019962.1 XP_016875451.1 19 3859
GenBank mRNA AB033034.2 - 12 2648
AK001821.1 - 2 203
AL359588.1 - 5 541
AM085438.1 - 19 3744
AY687932.1 - 19 3729
BC042615.1 - 7 1268
BC131787.1 - 19 3685 40126580
CR749871.1 - 5 541

View Full Product Details