Gene Transcripts

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
Chromosome Location
Chr. 10: 15778169 - 15860533 on Build GRCh38
UniGene ID
Hs.158870
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318330.1 NP_001305259.1 12 1253
NM_024948.3 NP_079224.1 13 1305
XM_005252600.2 XP_005252657.1 12 1199
XM_005252602.2 XP_005252659.1 11 1147
XM_006717508.2 XP_006717571.1 12 1201
XM_011519690.2 XP_011517992.1 13 1282
XM_011519691.2 XP_011517993.1 13 1282
XM_011519693.1 XP_011517995.1 13 1291
XM_011519694.1 XP_011517996.1 14 1346
XM_017016671.1 XP_016872160.1 12 1201
XM_017016672.1 XP_016872161.1 15 1438
XM_017016673.1 XP_016872162.1 13 1308
XM_017016674.1 XP_016872163.1 11 1149
XM_017016675.1 XP_016872164.1 11 1118
XM_017016676.1 XP_016872165.1 10 1066
GenBank mRNA AB014761.1 - 13 1071
AF063600.1 - 8 571
AF176916.1 - 12 1215
AK023459.1 - 13 1292
AL833873.1 - 13 1267
BC020605.1 - 12 1218
BC067799.1 - 13 1291 5278533
CR457315.1 - 13 1071

View Full Product Details