Gene Transcripts
- Gene Symbol
- CTSE
- Entrez Gene ID
- 1510
- Gene Name
- cathepsin E
- Gene Aliases
- CATE
- Chromosome Location
- Chr. 1: 206009264 - 206023909 on Build GRCh38
- UniGene ID
- Hs.644082
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317331.1 |
NP_001304260.1
|
4, 5 |
616 |
|
NM_001910.3 |
NP_001901.1
|
5, 6 |
773 |
|
NM_148964.2 |
NP_683865.1
|
5, 6 |
773 |
|
XM_011509244.1 |
XP_011507546.1
|
5, 6 |
778 |
|
XM_011509245.1 |
XP_011507547.1
|
5, 6 |
778 |
|
GenBank mRNA |
AJ250716.1 |
- |
5, 6 |
759 |
|
AJ250717.1 |
- |
5, 6 |
759 |
|
AK292057.1 |
- |
5, 6 |
703 |
|
AK298069.1 |
- |
4, 5 |
469 |
|
BC042537.1 |
- |
5, 6 |
740 |
5174814 |
J05036.1 |
- |
5, 6 |
704 |
|
View Full Product Details