Gene Transcripts

Gene Symbol
C19orf12
Entrez Gene ID
83636
Gene Name
chromosome 19 open reading frame 12
Gene Aliases
NBIA3, NBIA4, SPG43
Chromosome Location
Chr. 19: 29698886 - 29715789 on Build GRCh38
UniGene ID
Hs.529094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031726.3 NP_001026896.2 3 1428
NM_001256046.1 NP_001242975.1 4 1644
NM_001256047.1 NP_001242976.1 3 1505
NM_001282929.1 NP_001269858.1 2 1258
NM_001282930.1 NP_001269859.1 2 1529
NM_001282931.1 NP_001269860.1 4 1820
NM_031448.4 NP_113636.2 3 1699
GenBank mRNA AK057185.1 - 3 1396
AL162066.1 - 2 1203
BC004957.1 - 2 1189 3628073
BC009946.2 - 2 1189 4125477
BC017211.2 - 1 1089
BC063518.1 - 4 1323 4340755

View Full Product Details